Croconic acid disodium salt, 97%
Croconic acid disodium salt, 97%
Croconic acid disodium salt, 97%
Croconic acid disodium salt, 97%
Thermo Scientific Chemicals

Croconic acid disodium salt, 97%

CAS: 14379-00-1 | C5Na2O5 | 186.03 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
0.25 g
1 g
產品號碼 B22235.01
亦稱為 B22235-01
價格 (TWD)
-
申請報價
Quantity:
0.25 g
Request bulk or custom format
化學識別
CAS14379-00-1
IUPAC Namedisodium trioxocyclopent-1-ene-1,2-bis(olate)
Molecular FormulaC5Na2O5
InChI KeyOQXLFPHHAAAVKQ-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C1=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)Yellow to yellow-orange or yellow-green
Assay (ex Sulphated Ash)>96.0 to <104.0% (dry wt. basis)
FormPowder
Water Content (Karl Fischer Titration)<2.0%
Sulfated Ash73.3 - 79.4% (theory 76.35%, dry wt. basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only