4-(Trifluoromethyl)mandelic acid, 98%
4-(Trifluoromethyl)mandelic acid, 98%
4-(Trifluoromethyl)mandelic acid, 98%
4-(Trifluoromethyl)mandelic acid, 98%
Thermo Scientific Chemicals

4-(Trifluoromethyl)mandelic acid, 98%

CAS: 395-35-7 | C9H7F3O3 | 220.147 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22264.03
also known as B22264-03
Price (EUR)
28,30
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
28,30
Each
Add to cart
Chemical Identifiers
CAS395-35-7
IUPAC Name2-hydroxy-2-[4-(trifluoromethyl)phenyl]acetic acid
Molecular FormulaC9H7F3O3
InChI KeySDGXYUQKJPFLDG-UHFFFAOYNA-N
SMILESOC(C(O)=O)C1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)127.0-133.0?C
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only