Dichloroacetyl chloride, 97%
Dichloroacetyl chloride, 97%
Dichloroacetyl chloride, 97%
Thermo Scientific Chemicals

Dichloroacetyl chloride, 97%

CAS: 79-36-7 | C2HCl3O | 147.379 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B22296.36
also known as B22296-36
Price (TWD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS79-36-7
IUPAC Name2,2-dichloroacetyl chloride
Molecular FormulaC2HCl3O
InChI KeyFBCCMZVIWNDFMO-UHFFFAOYSA-N
SMILESClC(Cl)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4585-1.4635 @ 20?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only