3-Methyl-1-pentanol, 98%
3-Methyl-1-pentanol, 98%
Thermo Scientific Chemicals

3-Methyl-1-pentanol, 98%

Have Questions?
Catalog NumberQuantity
B22356.22
also known as B22356-22
100 g
Catalog number B22356.22
also known as B22356-22
Price (TWD)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS589-35-5
IUPAC Name3-methylpentan-1-ol
Molecular FormulaC6H14O
InChI KeyIWTBVKIGCDZRPL-UHFFFAOYNA-N
SMILESCCC(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4175-1.4215 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.