3,5-Dichlorobenzhydrazide, 97%
3,5-Dichlorobenzhydrazide, 97%
3,5-Dichlorobenzhydrazide, 97%
Thermo Scientific Chemicals

3,5-Dichlorobenzhydrazide, 97%

CAS: 62899-78-9 | C7H6Cl2N2O | 205.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22383.03
also known as B22383-03
Price (EUR)
31,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
31,00
Each
Chemical Identifiers
CAS62899-78-9
IUPAC Name3,5-dichlorobenzohydrazide
Molecular FormulaC7H6Cl2N2O
InChI KeyDNISXKBKTSHNKL-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC(Cl)=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)201.0-210.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only