2-(3-Phenylpropyl)pyridine, 99%
2-(3-Phenylpropyl)pyridine, 99%
2-(3-Phenylpropyl)pyridine, 99%
Thermo Scientific Chemicals

2-(3-Phenylpropyl)pyridine, 99%

CAS: 2110-18-1 | C14H15N | 197.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B22495.14
also known as B22495-14
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS2110-18-1
IUPAC Name2-(3-phenylpropyl)pyridine
Molecular FormulaC14H15N
InChI KeyJJJPNTQYUJPWGQ-UHFFFAOYSA-N
SMILESC(CC1=CC=CC=C1)CC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.5585-1.5615 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only