2-Mercapto-1,3,4-thiadiazole, 98%
2-Mercapto-1,3,4-thiadiazole, 98%
2-Mercapto-1,3,4-thiadiazole, 98%
Thermo Scientific Chemicals

2-Mercapto-1,3,4-thiadiazole, 98%

CAS: 18686-82-3 | C2H2N2S2 | 118.172 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B22556.09
also known as B22556-09
Price (EUR)
34,10
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
34,10
Each
Chemical Identifiers
CAS18686-82-3
IUPAC Name2,3-dihydro-1,3,4-thiadiazole-2-thione
Molecular FormulaC2H2N2S2
InChI KeyJLAMDELLBBZOOX-UHFFFAOYSA-N
SMILESS=C1NN=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)141-147?C
FormCrystals or crystalline powder
2-Mercapto-1,3,4-thiadiazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercapto-1,3,4-thiadiazole is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Nisha Aggarwal; Rajesh Kumar; Prem Dureja; Jitender Mohan Khurana. Synthesis of novel nalidixic acid-based 1,3,4-thiadiazole and 1,3,4-oxadiazole derivatives as potent antibacterial agents. Chemical Biology & Drug Design. 2012, 79,(4), 384-397.