4-n-Dodecyloxybenzoic acid, 98%
4-n-Dodecyloxybenzoic acid, 98%
4-n-Dodecyloxybenzoic acid, 98%
Thermo Scientific Chemicals

4-n-Dodecyloxybenzoic acid, 98%

CAS: 2312-15-4 | C19H30O3 | 306.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B22689.14
also known as B22689-14
Price (EUR)
130,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
130,00
Each
Chemical Identifiers
CAS2312-15-4
IUPAC Name4-(dodecyloxy)benzoic acid
Molecular FormulaC19H30O3
InChI KeyALQLYJHDBAKLBB-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCOC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5%
FormPowder
4-n-Dodecyloxybenzoic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Dodecyloxybenzoic acid is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water. Soluble in N,N-Dimethylformamide.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. H.N.Shreenivasa Murthya; B.K.Sadashiva. Influence of a fluorine substituent on the mesomorphic properties of unsymmetrical five-ring bent-core compounds. J. Mater. Chem. 2004, 14,(18), 2813-2821.
  2. A.Biering; D.Demus; G.W.Gray; H.Sackmann. The classification of the liquid crystalline modificatons in some homologous series. Molecular Crystals and Liquid Crystals. 1974, 28,(3-4), 275-292.