5-Chlorothiophene-2-carboxaldehyde, 97%
5-Chlorothiophene-2-carboxaldehyde, 97%
5-Chlorothiophene-2-carboxaldehyde, 97%
Thermo Scientific Chemicals

5-Chlorothiophene-2-carboxaldehyde, 97%

CAS: 7283-96-7 | C5H3ClOS | 146.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22833.06
also known as B22833-06
Price (EUR)
120,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
120,00
Each
Chemical Identifiers
CAS7283-96-7
IUPAC Name5-chlorothiophene-2-carbaldehyde
Molecular FormulaC5H3ClOS
InChI KeyVWYFITBWBRVBSW-UHFFFAOYSA-N
SMILESClC1=CC=C(S1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Appearance (Color)Clear yellow to orange to red
Refractive Index1.6040-1.6090 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only