(Di-n-dodecyl)dimethylammonium bromide is used as a phase transfer catalyst. It is also used as floor disinfectant. Further, it acts as surface-active agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(Di-n-dodecyl)dimethylammonium bromide is used as a phase transfer catalyst. It is also used as floor disinfectant. Further, it acts as surface-active agent.
Solubility
Soluble in hot and cold water.
Notes
Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- Wu, H.; Wang, X.; Qiao, M.; Zhang, H.; Jin, X.; Fan, S. Enhancing sensitivity of hemoglobin-based electrochemical biosensor by using protein conformational intermediate. Sens. Actuators, B 2015, 221, 694-699.
- Ma, W.; Xu, A.; Ying, J.; Li, B.; Jin, Y. Biodegradable Core-Shell Copolymer-Phospholipid Nanoparticles for Combination Chemotherapy: An In Vitro Study. J. Biomed. Nanotechnol. 2015, 11 (7), 1193-1200.