Thermo Scientific Chemicals

3-Dimethylaminophenol, 97+%, Thermo Scientific Chemicals

Catalog number: B23067.14
25 g, Each
Thermo Scientific Chemicals

3-Dimethylaminophenol, 97+%, Thermo Scientific Chemicals

Catalog number: B23067.14
25 g, Each
Quantity
Catalog number: B23067.14
also known as B23067-14
Price (USD)
Price: 112.00
Online price: 96.65
Your price:
Quantity
-

Chemical Identifiers

CAS
99-07-0
Appearance (Color)
Grey to brown to dark brown to purple to dark purple
Form
Powder
Assay (GC)
≥97.0%
Identification (FTIR)
Conforms

Description

3-Dimethylaminophenol can be used as neostigmine bromide intermediate and dye intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Dimethylaminophenol can be used as neostigmine bromide intermediate and dye intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from oxidizing agents, light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text