3-(2-Aminoethylamino)propylmethyldimethoxysilane, 97%
3-(2-Aminoethylamino)propylmethyldimethoxysilane, 97%
3-(2-Aminoethylamino)propylmethyldimethoxysilane, 97%
Thermo Scientific Chemicals

3-(2-Aminoethylamino)propylmethyldimethoxysilane, 97%

CAS: 3069-29-2 | C8H22N2O2Si | 206.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B23159.22
also known as B23159-22
Price (USD)
62.65
Online Exclusive
69.60
Save 6.95 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
62.65
Online Exclusive
69.60
Save 6.95 (10%)
Each
Chemical Identifiers
CAS3069-29-2
IUPAC Name(2-aminoethyl)({3-[dimethoxy(methyl)silyl]propyl})amine
Molecular FormulaC8H22N2O2Si
InChI KeyMQWFLKHKWJMCEN-UHFFFAOYSA-N
SMILESCO[Si](C)(CCCNCCN)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4465-1.4515 @ 20?C
Assay (GC)≥96.0%
FormLiquid
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only