Bathophenanthrolinedisulfonic acid disodium salt hydrate, 98%
Bathophenanthrolinedisulfonic acid disodium salt hydrate, 98%
Bathophenanthrolinedisulfonic acid disodium salt hydrate, 98%
Thermo Scientific Chemicals

Bathophenanthrolinedisulfonic acid disodium salt hydrate, 98%

CAS: 52746-49-3 | C24H14N2Na2O6S2 | 536.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number B23244.MD
also known as B23244-MD
Price (TWD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS52746-49-3
IUPAC Namedisodium 4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzene-1-sulfonate
Molecular FormulaC24H14N2Na2O6S2
InChI KeyPCNDSIWXTYFWIA-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=CC=NC2=C1C=CC1=C2N=CC=C1C1=CC=C(C=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pink to orange
FormPowder
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0% (dry wt. basis)
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)2 to 12%
Bathophenanthrolinedisulfonic acid disodium salt hydrate is used as colorimetric reagent for detection of iron. It is widely used for aqueous iron determination without the usage of solvent for extraction. It is used as precursor in the preparation of anionic Europium(III) disulfonated bathophenanthroline tris (dibenzoylmethanate) complex and ruthenium II tris (bathophenanthroline disulfonate). It plays a vital role in the catyltic oxidation of 2-hexanol by making it a water soluble complex with palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bathophenanthrolinedisulfonic acid disodium salt hydrate is used as colorimetric reagent for detection of iron. It is widely used for aqueous iron determination without the usage of solvent for extraction. It is used as precursor in the preparation of anionic Europium(III) disulfonated bathophenanthroline tris (dibenzoylmethanate) complex and ruthenium II tris (bathophenanthroline disulfonate). It plays a vital role in the catyltic oxidation of 2-hexanol by making it a water soluble complex with palladium.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Xu, S. S.; Lin, S. Z.; Lai, Z. X. Cadmium impairs iron homeostasis in Arabidopsis thaliana by increasing the polysaccharide contents and the iron-binding capacity of root cell walls. Plant and Soil 2015, 392 (1), 71-85.
  2. Rydzek, G.; Ji, Q.; Li, M.; Schaaf, P.; Hill, J. P.; Boulmedais, F.; Ariga, K. Electrochemical nanoarchitectonics and layer-by-layer assembly: From basics to future. Nano Today 2015, 10 (2), 138-167.