Thermo Scientific Chemicals

Acenaphthenequinone, 95%, Thermo Scientific Chemicals

Catalog number: B23466.14
25 g, Each
Thermo Scientific Chemicals

Acenaphthenequinone, 95%, Thermo Scientific Chemicals

Catalog number: B23466.14
25 g, Each
Quantity
Catalog number: B23466.14
also known as B23466-14
Price (USD)
Price: 66.10
Online price: 56.65
Your price:
Quantity
-

Chemical Identifiers

CAS
82-86-0
IUPAC Name
1,2-dihydroacenaphthylene-1,2-dione
Molecular Formula
C12H6O2
InChI Key
AFPRJLBZLPBTPZ-UHFFFAOYSA-N
SMILES
O=C1C(=O)C2=C3C1=CC=CC3=CC=C2
Appearance (Color)
Yellow
Assay (HPLC)
≥94.0%
Melting Point (clear melt)
252-263?C
Form
Powder
Identification (FTIR)
Conforms

Description

Acenaphthenequinone is used in the preparation of acetylcholinesterase inhibitory agents. It is also used in the preparation of turn-on sensors for cysteine/homocysteine and its application in bioimaging. Further, it is used as an intermediate in manufacturing of dyes and pharmaceuticals. In addition to this, it is used in chemical research as a drug and therapeutic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acenaphthenequinone is used in the preparation of acetylcholinesterase inhibitory agents. It is also used in the preparation of turn-on sensors for cysteine/homocysteine and its application in bioimaging. Further, it is used as an intermediate in manufacturing of dyes and pharmaceuticals. In addition to this, it is used in chemical research as a drug and therapeutic agent.

Solubility
Soluble in chloroform and alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text