Buffers for RNA Transctription and Post-Transcriptional Modifications
Thermo Scientific™

Buffers for RNA Transctription and Post-Transcriptional Modifications

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidadNombre del producto químico o materialConcentración
B2348AF1000 μLTampón de poli(A) polimerasa10X
F8156AF200 μLTampón de transcripción10X
B23AF800 μLSolución tampón10X
B1299AF200 μLTampón de reacción de ARN ligasa de T410X
B3277AF200 μLTampón de reacción de ARN ligasa 2 de T410X
Número de catálogo B2348AF
Precio (USD)
-
Cantidad:
1000 μL
Nombre del producto químico o material:
Tampón de poli(A) polimerasa
Concentración:
10X

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Features

  • Convenient stand-alone enzymes and buffers for workflow optimization
  • No animal-origin components in final product composition
  • Reliable and reproducible results with rigorous quality testing
  • Scalable and available as OEM products

Applications

  • CRISPR/Cas-based genome editing
  • Gene expression
  • Generation of pluripotent cells

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.

Especificaciones
Nombre del producto químico o materialTampón de poli(A) polimerasa
IncluyePoly(A) Polymerase Buffer, 1000 μL

Store at -20°C.
Concentración10X
Para utilizar con (aplicación)Polyadenylation of RNA
Forma físicaLíquido
Cantidad1000 μL
Unit SizeEach