2-Chloro-4-nitrotoluene, 98%
2-Chloro-4-nitrotoluene, 98%
2-Chloro-4-nitrotoluene, 98%
Thermo Scientific Chemicals

2-Chloro-4-nitrotoluene, 98%

CAS: 121-86-8 | C7H6ClNO2 | 171.58 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
產品號碼 B23542.22
亦稱為 B23542-22
價格 (TWD)
-
Quantity:
100 g
Request bulk or custom format
化學識別
CAS121-86-8
IUPAC Name2-chloro-1-methyl-4-nitrobenzene
Molecular FormulaC7H6ClNO2
InChI KeyLLYXJBROWQDVMI-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1Cl)[N+]([O-])=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to pale yellow to pale orange
Assay (GC)≥97.5%
Melting Point (clear melt)60.0-67.0?C
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only