Thermo Scientific Chemicals

Di-n-butyl sulfoxide, 97%, Thermo Scientific Chemicals

Catalog number: B23672.06
5 g, Each
Thermo Scientific Chemicals

Di-n-butyl sulfoxide, 97%, Thermo Scientific Chemicals

Catalog number: B23672.06
5 g, Each
Quantity
Catalog number: B23672.06
also known as B23672-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
2168-93-6
IUPAC Name
1-(butane-1-sulfinyl)butane
Molecular Formula
C8H18OS
InChI Key
LOWMYOWHQMKBTM-UHFFFAOYSA-N
SMILES
CCCCS(=O)CCCC
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
29.0-38.0?C
Appearance (Color)
White
Assay (GC)
≥96.0%

Description

It is used as an active pharmaceutical intermediate and in the extraction of thorium(IV) from nitric acid solutions in xylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate and in the extraction of thorium(IV) from nitric acid solutions in xylene.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Protect from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text