Thermo Scientific Chemicals

3-Chlorostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: B23806.06
5 g, Each
Thermo Scientific Chemicals

3-Chlorostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: B23806.06
5 g, Each
Quantity
Catalog number: B23806.06
also known as B23806-06
Price (USD)
Price: 92.10
Online price: 78.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2039-85-2
IUPAC Name
1-chloro-3-ethenylbenzene
Molecular Formula
C8H7Cl
InChI Key
BOVQCIDBZXNFEJ-UHFFFAOYSA-N
SMILES
ClC1=CC=CC(C=C)=C1
Refractive Index
1.5600-1.5640 @ 20°C
Form
Liquid
Solution Test
10% in diethyl ether: clear solution
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to pale yellow

Description

It has been identified as a potential candidate for an electron resist to be used in direct device fabrication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been identified as a potential candidate for an electron resist to be used in direct device fabrication.

Solubility
Not miscible or difficult to mix in water. Soluble in ether, and ethanol

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text