Thermo Scientific Chemicals

Heptanal, 97 %, Thermo Scientific Chemicals

Número de catálogo: B23830.AU
1000 mL, Each
Thermo Scientific Chemicals

Heptanal, 97 %, Thermo Scientific Chemicals

Número de catálogo: B23830.AU
1000 mL, Each
Cantidad
Número de catálogo: B23830.AU
también denominado B23830-AU
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
111-71-7
IUPAC Name
heptanal
Molecular Formula
C7H14O
InChI Key
FXHGMKSSBGDXIY-UHFFFAOYSA-N
SMILES
CCCCCCC=O
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Free acid (titration)
≤2.0%
Identification (FTIR)
Conforms
Refractive Index
1.4100-1.4150 @ 20?C

Descripción

Heptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El heptanal se utiliza como intermediario de síntesis en la industria de las fragancias y los sabores. Es el precursor del 1-heptanol, el heptanoato de etilo y ciertos lubricantes.

Solubilidad
Soluble en agua en una proporción de 1,3 g/l.

Notas
Higroscópico. Almacenar lejos de agentes oxidantes y del aire.
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 3
    Skin Corrosion/Irritation
    Category 2
    Chronic aquatic toxicity
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H412 - Harmful to aquatic life with long lasting effects

    Physical Hazards

    H226 - Flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P280 - Wear protective gloves/protective clothing

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P332 + P313 - If skin irritation occurs: Get medical advice/attention