2,2,2-Trifluoro-N-methylacetamide, 98%
2,2,2-Trifluoro-N-methylacetamide, 98%
2,2,2-Trifluoro-N-methylacetamide, 98%
Thermo Scientific Chemicals

2,2,2-Trifluoro-N-methylacetamide, 98%

CAS: 815-06-5 | C3H4F3NO | 127.066 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B23851.14
also known as B23851-14
Price (EUR)
126,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
126,00
Each
Chemical Identifiers
CAS815-06-5
IUPAC Name2,2,2-trifluoro-N-methylacetamide
Molecular FormulaC3H4F3NO
InChI KeyIQNHBUQSOSYAJU-UHFFFAOYSA-N
SMILESCNC(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)44.0-52.0?C
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only