Thermo Scientific Chemicals

m-Toluoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: B23914.36
500 g, Each
Thermo Scientific Chemicals

m-Toluoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: B23914.36
500 g, Each
Quantity
Catalog number: B23914.36
also known as B23914-36
Price (USD)
Price: 270.00
Online price: 229.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1711-06-4
IUPAC Name
3-methylbenzoyl chloride
Molecular Formula
C8H7ClO
InChI Key
YHOYYHYBFSYOSQ-UHFFFAOYSA-N
SMILES
CC1=CC=CC(=C1)C(Cl)=O
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Refractive Index
1.5475-1.5505 @ 20°C

Description

m-Toluoyl chloride has been used to derivatize the C1-C4 aliphatic amines for their determination in air by HPLC. It has been used in determination of atmospheric ammonia by denuder-sampling and HPLC-UV detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Toluoyl chloride has been used to derivatize the C1-C4 aliphatic amines for their determination in air by HPLC. It has been used in determination of atmospheric ammonia by denuder-sampling and HPLC-UV detection.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, active metals, amines and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text