Di-n-butyltin bis(2,4-pentanedionate), 95%
Di-n-butyltin bis(2,4-pentanedionate), 95%
Di-n-butyltin bis(2,4-pentanedionate), 95%
Thermo Scientific Chemicals

Di-n-butyltin bis(2,4-pentanedionate), 95%

CAS: 22673-19-4 | C18H32O4Sn
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B23921.0B
also known as B23921-0B
Price (USD)
-
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS22673-19-4
IUPAC Nametin(4+) bis(2,4-dioxopentan-3-ide) bis(pentan-1-ide)
Molecular FormulaC20H36O4Sn
InChI KeyVJLXWWRKGIBBSL-UHFFFAOYSA-N
SMILES[Sn+4].CCCC[CH2-].CCCC[CH2-].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormViscous liquid
Refractive Index1.5100-1.5230 @ 20°C
Elemental AnalysisTin, Sn: 26-28% (from Vendor's Certificate of Analysis, Theory: 27.53%)
Appearance (Color)Clear yellow to orange
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only