Dichloromethylphenylsilane, 98%
Dichloromethylphenylsilane, 98%
Dichloromethylphenylsilane, 98%
Thermo Scientific Chemicals

Dichloromethylphenylsilane, 98%

CAS: 149-74-6 | C7H8Cl2Si | 191.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B23999.36
also known as B23999-36
Price (EUR)
228,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
228,00
Each
Chemical Identifiers
CAS149-74-6
IUPAC Namedichloro(methyl)phenylsilane
Molecular FormulaC7H8Cl2Si
InChI KeyGNEPOXWQWFSSOU-UHFFFAOYSA-N
SMILESC[Si](Cl)(Cl)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale pink
Assay (GC)≥97.5%
Refractive Index1.5170-1.5210 @ 20?C
Identification (FTIR)Conforms
Dichloromethylphenylsilane is used as a monomer (‘building block’) in the production of silicone polymers or silicone resins. Silicone polymers may be oils, greases and rubbers. It is also used as an intermediate (starting material) in the production of other organosilicon substances. It is a bifunctional reagent for the preparation of methylphenylsilyl derivatives of diols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichloromethylphenylsilane is used as a monomer (‘building block’) in the production of silicone polymers or silicone resins. Silicone polymers may be oils, greases and rubbers. It is also used as an intermediate (starting material) in the production of other organosilicon substances. It is a bifunctional reagent for the preparation of methylphenylsilyl derivatives of diols.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is sensitive to moisture. Protect from humidity and water.
RUO – Research Use Only

General References:

  1. Simon J. Holder.; Mariliz Achilleos. An oligosilane initiator for the Wurtz-type polymerization of dichloromethylphenylsilane. Journal of Organometallic Chemistry. 2008, 693 (10), 1938-1944.
  2. Atsutaka. Kunai.; Toshihiro. Kawakami.; Eiji. Toyoda.; Mitsuo. Ishikawa. Electrochemistry of organosilicon compounds. 2. Synthesis of polysilane oligomers by a copper electrode system. J. Org. Chem. 1991, 10 (6), 2001-2003.
  3. Bifunctional reagent for the preparation of methylphenylsilyl derivatives of diols.