Ethyl 3-ethoxypropionate is used in the synthesis of phenols and selective inhibitors of cyclin-dependant kinase 4/6 for novel cancer therapies. It is also used as a solvent to prepare polymers. Further, it is used in paints and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El 3-etoxipropionato de etilo se utiliza en la síntesis de fenoles e inhibidores selectivos de la quinasa dependiente de ciclina 4/6 para nuevas terapias contra el cáncer. También se utiliza como disolvente para preparar polímeros. Además, se utiliza en pinturas y recubrimientos.
Solubilidad
Miscible en la mayoría de los disolventes orgánicos. Ligeramente miscible en agua fría.
Notas
Incompatible con agentes oxidantes fuertes, metales y ácidos fuertes.
RUO – Research Use Only
General References:
- Rabnawaz, M.; Liu, G.; Hu, H. Fluorine-Free Anti-Smudge Polyurethane Coatings. Angew. Chem. Int. Ed. 2015, 127 (43), 12913-12918.
- Wang, C. F.; Kuo, S. W.; Lin, C. H.; Chen, H. G.; Liao, C. S.; Hung, P. R. Benzoxazine as a reactive noncovalent dispersant for carbon nanotubes. RSC Adv. 2014, 4 (68), 36012-36016.