5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4in.-disulfonic acid monosodium salt hydrate, 97%
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4in.-disulfonic acid monosodium salt hydrate, 97%
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4in.-disulfonic acid monosodium salt hydrate, 97%
Thermo Scientific Chemicals

5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4in.-disulfonic acid monosodium salt hydrate, 97%

CAS: 1266615-85-3 | C20H15N4NaO7S2 | 510.471 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24066.06
also known as B24066-06
Price (USD)
147.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
147.00
Each
Chemical Identifiers
CAS1266615-85-3
IUPAC Namesodium 4-[3-(pyridin-2-yl)-6-(4-sulfophenyl)-1,2,4-triazin-5-yl]benzene-1-sulfonate
Molecular FormulaC20H13N4NaO6S2
InChI KeyZGVNYCXXBQPDPQ-UHFFFAOYSA-M
SMILES[Na+].OS(=O)(=O)C1=CC=C(C=C1)C1=C(N=C(N=N1)C1=CC=CC=N1)C1=CC=C(C=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0% (dry wt. basis)
FormCrystals or powder or crystalline powder
Appearance (Color)Yellow to yellow-green
Water Content (Karl Fischer Titration)≤10.0%
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4-disulfonic acid monosodium salt hydrate is used in the accurate determination of sub microgram levels of serum iron. (i.e.. reagent for the spectrophotometric determination of Fe).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4″-disulfonic acid monosodium salt hydrate is used in the accurate determination of sub microgram levels of serum iron. (i.e.. reagent for the spectrophotometric determination of Fe).

Solubility
Soluble in water (partly).

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. D Plaut; E Miller; J MacQueen; R Blanchard. Direct and indirect assays of serum iron with a sensitive new color reagent. The American journal of medical technology. 1972, 38 (3), 82-89.
  2. F Ceriotti; G Ceriotti. Improved direct specific determination of serum iron and total iron-binding capacity. Clinical Chemistry. 1980, 26 (2), 327-331.