1,3-Dibromobutane, 98%
1,3-Dibromobutane, 98%
1,3-Dibromobutane, 98%
Thermo Scientific Chemicals

1,3-Dibromobutane, 98%

CAS: 107-80-2 | C4H8Br2 | 215.916 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B24123.22
also known as B24123-22
Price (EUR)
65,30
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
65,30
Each
Chemical Identifiers
CAS107-80-2
IUPAC Name1,3-dibromobutane
Molecular FormulaC4H8Br2
InChI KeyXZNGUVQDFJHPLU-UHFFFAOYNA-N
SMILESCC(Br)CCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.5075-1.5115 @ 20?C
Identification (FTIR)Conforms
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only