1-Octylamine, 99%
1-Octylamine, 99%
1-Octylamine, 99%
Thermo Scientific Chemicals

1-Octylamine, 99%

CAS: 111-86-4 | C8H19N | 129.247 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B24193.22
also known as B24193-22
Price (EUR)
25,10
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
25,10
Each
Chemical Identifiers
CAS111-86-4
IUPAC Nameoctan-1-amine
Molecular FormulaC8H19N
InChI KeyIOQPZZOEVPZRBK-UHFFFAOYSA-N
SMILESCCCCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
FormLiquid
Refractive Index1.4275-1.4305 @ 20?C
1-Octylamine is used as a precursor to prepare chemicals. It finds application in functional fluids and as a laboratory reagent. Further, it is used as a reactant in the preparation of 2H-indazoles and 1H-indazolones, which are used as myeloperoxidase(MPO) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octylamine is used as a precursor to prepare chemicals. It finds application in functional fluids and as a laboratory reagent. Further, it is used as a reactant in the preparation of 2H-indazoles and 1H-indazolones, which are used as myeloperoxidase(MPO) inhibitors.

Solubility
Miscible with ethanol, benzene and ethyl acetate. Slightly miscible with water.

Notes
Incompatible with acids, strong oxidizing agents, acid anhydrides and acid chlorides.
RUO – Research Use Only

General References:

  1. Firmansyah, D.; Banasiewicz, M.; Gryko, D. T. Vertically-expanded imidazo[1,2-a]pyridines and imidazo[1,5-a]pyridinevia dehydrogenative coupling. Org. Biomol. Chem. 2015, 13 (5), 1367-1374.
  2. Liu, Z.; Chiang, C. Y.; Li, W.; Zhou, W. The role of surface hydrolysis of ferricyanide anions in crystal growth of snowflake-shaped alpha-Fe2O3. Chem. Commun. 2015, 51 (45), 9350-9353.