1-Iodoheptane, 98%, stab. with copper
1-Iodoheptane, 98%, stab. with copper
1-Iodoheptane, 98%, stab. with copper
Thermo Scientific Chemicals

1-Iodoheptane, 98%, stab. with copper

CAS: 4282-40-0 | C7H15I | 226.101 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number B24206.30
also known as B24206-30
Price (TWD)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS4282-40-0
IUPAC Name1-iodoheptane
Molecular FormulaC7H15I
InChI KeyLMHCYRULPLGEEZ-UHFFFAOYSA-N
SMILESCCCCCCCI
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
CommentCopper powder used as stabilizer
Refractive Index1.4880-1.4920 @ 20?C
Appearance (Color)Clear colorless to pale pink
Assay (GC)>97.5%
1-Iodoheptane is used in the functionalization of symmetrical star-shaped molecules with carborane clusters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodoheptane is used in the functionalization of symmetrical star-shaped molecules with carborane clusters.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. It is sensitive to light. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Mikhail F. Bolotnikov.; Yurij A. Neruchev. Temperature Dependence of the Thermophysical Properties of 1-Chlorohexane, 1-Iodohexane, 1-Iodoheptane, and 1-Chlorononane at Saturation Condition. Chem. Eng. Data. 2004, 49 (2), 202-207.
  2. Mikhail F. Bolotnikov.; Yurij A. Neruchev. Relative Permittivity for 1-Chlorohexane, 1-Iodohexane, 1-Iodoheptane, and 1-Chlorononane from (293.15 to 373.15) K and Hexane + 1-Chlorohexane from (293.15 to 333.15) K. Chem. Eng. Data. 2004, 49 (1), 895-898.