3-Ethoxyphenol, 98%
3-Ethoxyphenol, 98%
3-Ethoxyphenol, 98%
Thermo Scientific Chemicals

3-Ethoxyphenol, 98%

CAS: 621-34-1 | C8H10O2 | 138.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B24266.06
also known as B24266-06
Price (USD)
57.65
Special offer
Online exclusive
Ends: 30-Jun-2026
68.00
Save 10.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
57.65
Special offer
Online exclusive
Ends: 30-Jun-2026
68.00
Save 10.35 (15%)
Each
Chemical Identifiers
CAS621-34-1
IUPAC Name3-ethoxyphenol
Molecular FormulaC8H10O2
InChI KeyVBIKLMJHBGFTPV-UHFFFAOYSA-N
SMILESCCOC1=CC=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear pale yellow to yellow to orange or pink
Assay (GC)≥97.5%
Refractive Index1.5390-1.5430 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only