2-Bromo-4'-phenylacetophenone, 97%
2-Bromo-4'-phenylacetophenone, 97%
2-Bromo-4'-phenylacetophenone, 97%
2-Bromo-4'-phenylacetophenone, 97%
Thermo Scientific Chemicals

2-Bromo-4'-phenylacetophenone, 97%

CAS: 135-73-9 | C14H11BrO | 275.145 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B24298.14
also known as B24298-14
Price (EUR)
84,70
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
84,70
Each
Add to cart
Chemical Identifiers
CAS135-73-9
IUPAC Name1-{[1,1'-biphenyl]-4-yl}-2-bromoethan-1-one
Molecular FormulaC14H11BrO
InChI KeyKGHGZRVXCKCJGX-UHFFFAOYSA-N
SMILESBrCC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown
FormPowder
Assay (GC)≥96.0%
Melting Point (clear melt)122.5-128.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only