Congo Red, indicator grade
Congo Red, indicator grade
Congo Red, indicator grade
Thermo Scientific Chemicals

Congo Red, indicator grade

Have Questions?
Catalog NumberQuantity
B24310.1425g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B24310.14
Price (EUR)
-
Quantity:
25g
Request bulk or custom format
Chemical Identifiers
CAS573-58-0
IUPAC Namedisodium 4-amino-3-(2-{4'-[2-(1-amino-4-sulfonatonaphthalen-2-yl)diazen-1-yl]-[1,1'-biphenyl]-4-yl}diazen-1-yl)naphthalene-1-sulfonate
Molecular FormulaC32H22N6Na2O6S2
InChI KeyIQFVPQOLBLOTPF-UHFFFAOYSA-L
SMILES[Na+].[Na+].NC1=C2C=CC=CC2=C(C=C1N=NC1=CC=C(C=C1)C1=CC=C(C=C1)N=NC1=CC(=C2C=CC=CC2=C1N)S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Specific Absorbance (E 1%/1cm)>500 @ lambda-max (pH 7)
Loss on Drying≤5.0%
Solution Test0.1% in water: clear
lambda-max (UV-Vis)490-500nm @ pH 7
View more
Congo red is useful for staining and visual detection of amyloid deposits. It inhibits the accumulation of the scrapie-associated, protease-resistant isoform of protein PrP without affecting the metabolism of the normal isoform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Congo red is useful for staining and visual detection of amyloid deposits. It inhibits the accumulation of the scrapie-associated, protease-resistant isoform of protein PrP without affecting the metabolism of the normal isoform.

Solubility
Soluble in water 25 g/L.

Notes
Store away from oxidizing agents, acids, reducing agents.
RUO – Research Use Only
Young Hye Kim; Se Hoon Choi; Carla D’Avanzo; Matthias Hebisch; Christopher Sliwinski; Enjana Bylykbashi; Kevin J Washicosky; Justin B Klee; Oliver Brüstle; Rudolph E Tanzi; Doo Yeon Kim. A 3D human neural cell culture system for modeling Alzheimer’s disease. Nature Protocols. 2015, 10 (7), 985-1006.

D P Penney; J M Powers; M Frank; C Willis; C Churukian. Analysis and testing of biological stains--the Biological Stain Commission Procedures. Biotechnic and Histochemistry. 2002, 77 (5-6), 237-275.