3-Chloropyridine, 99%
3-Chloropyridine, 99%
3-Chloropyridine, 99%
Thermo Scientific Chemicals

3-Chloropyridine, 99%

CAS: 626-60-8 | C5H4ClN | 113.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B24316.18
also known as B24316-18
Price (TWD)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS626-60-8
IUPAC Name3-chloropyridine
Molecular FormulaC5H4ClN
InChI KeyPWRBCZZQRRPXAB-UHFFFAOYSA-N
SMILESClC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to yellow
Assay (GC)≥98.5%
FormLiquid
Refractive Index1.5310-1.5350 @ 20°C (non-U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
3-Chloropyridine is used as a pharmaceutical intermediate. It can be used to produce 3-chloro-pyridine-1-oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropyridine is used as a pharmaceutical intermediate. It can be used to produce 3-chloro-pyridine-1-oxide.

Solubility
Soluble in water 10 g/L.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. S. M. Angel and D. D. Archibald. Near-Infrared Surface-Enhanced Raman Spectra of 3-Picoline and 3-Chloropyridine on a Copper Electrode. Applied Spectroscopy. 1989, 43 (6), 1097-1099.
  2. José M. Moreno; Maria A. Aramendía; Alberto Marinas; José M. Marinas; Francisco J. Urbano. Hydrodechlorination of 3-chloropyridine and chlorobenzene in methanol solution over alkali-modified zirconia-supported palladium catalysts. Applied Catalysis B: Environmental. 2005, 59 (3-4), 275-283.
  3. With n-BuLi and TMEDA at -60 °, lithiation occurs predominantly at the 2-position, with some 4-lithiation: Tetrahedron, 39, 2009 (1983). With LDA at -60 °, exclusive 4-lithiation occurs: J. Org. Chem., 47, 2633 (1982); Heterocycles, 35, 151 (1993).