Thermo Scientific Chemicals

4-Bromo-2,2-difluoro-1,3-benzodioxole, 97%, Thermo Scientific Chemicals

Catalog number: B24389.06
5 g, Each
Thermo Scientific Chemicals

4-Bromo-2,2-difluoro-1,3-benzodioxole, 97%, Thermo Scientific Chemicals

Catalog number: B24389.06
5 g, Each
Quantity
Catalog number: B24389.06
also known as B24389-06
Price (USD)
Price: 206.00
Online price: 178.65
Your price:
Quantity
-

Chemical Identifiers

CAS
144584-66-7
IUPAC Name
4-bromo-2,2-difluoro-2H-1,3-benzodioxole
Molecular Formula
C7H3BrF2O2
InChI Key
LSZYHXNOLVSZHH-UHFFFAOYSA-N
SMILES
FC1(F)OC2=CC=CC(Br)=C2O1
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to brown
Refractive Index
1.4950-1.5000 @ 20°C
Form
Liquid

Description

4-Bromo-2,2-difluoro-1,3-benzodioxole is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,2-difluoro-1,3-benzodioxole is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
It is unstable under normal temperature. storage temperature is 2 - 4°C. Incompatible material is oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text