Thermo Scientific Chemicals

3-Chlorobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B24444.03
1 g, Each
Thermo Scientific Chemicals

3-Chlorobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: B24444.03
1 g, Each
Quantity
Catalog number: B24444.03
also known as B24444-03
Price (USD)
Price: 24.40
Online price: 20.65
Your price:
Quantity
-

Chemical Identifiers

CAS
63503-60-6
IUPAC Name
(3-chlorophenyl)boronic acid
Molecular Formula
C6H6BClO2
InChI Key
SDEAGACSNFSZCU-UHFFFAOYSA-N
SMILES
OB(O)C1=CC(Cl)=CC=C1
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0% (non-U.S. specification)
Assay (HPLC)
≥96.0%
Comment
Specification differs for U.S. and non-U.S. material where indicated

Description

It is employed as a reactant involved in 1,4-conjugate addition reactions with ethenesulfonamides to form arylethanesulfonamides, Suzuki-Miyaura reactions with dibromotrifluoromethylbenzene, cross-coupling reactions with diazoesters3 or potassium cyanate. It is involved in the synthesis of biarylketones and phthalides and synthesis of inhibitors including PDE4 inhibitors6 among others.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a reactant involved in 1,4-conjugate addition reactions with ethenesulfonamides to form arylethanesulfonamides, Suzuki-Miyaura reactions with dibromotrifluoromethylbenzene, cross-coupling reactions with diazoesters3 or potassium cyanate. It is involved in the synthesis of biarylketones and phthalides and synthesis of inhibitors including PDE4 inhibitors6 among others.

Solubility
Slightly soluble in water. soluble in ether, tetrahydrofuran,, dimethyl sulfoxide, dimethyl formamide, methanol.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text