Thermo Scientific Chemicals

Isopropyl salicylate, 99%, Thermo Scientific Chemicals

Catalog number: B24471.22
100 g, Each
Thermo Scientific Chemicals

Isopropyl salicylate, 99%, Thermo Scientific Chemicals

Catalog number: B24471.22
100 g, Each
Quantity
Catalog number: B24471.22
also known as B24471-22
Price (USD)
Price: 195.00
Online price: 168.65
Your price:
Quantity
-

Chemical Identifiers

CAS
607-85-2
IUPAC Name
propan-2-yl 2-hydroxybenzoate
Molecular Formula
C10H12O3
InChI Key
YEULQIJMIOWCHB-UHFFFAOYSA-N
SMILES
CC(C)OC(=O)C1=CC=CC=C1O
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Form
Liquid
Refractive Index
1.5085-1.5115 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text