5-Chloro-2(3H)-benzoxazolone, 99%, Thermo Scientific Chemicals
5-Chloro-2(3H)-benzoxazolone, 99%, Thermo Scientific Chemicals
5-Chloro-2(3H)-benzoxazolone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Chloro-2(3H)-benzoxazolone, 99%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
B24507.18
also known as B24507-18
50 g
Catalog number B24507.18
also known as B24507-18
Price (EUR)
44,60
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
44,60
Each
Chemical Identifiers
CAS95-25-0
IUPAC Name5-chloro-2,3-dihydro-1,3-benzoxazol-2-one
Molecular FormulaC7H4ClNO2
InChI KeyTZFWDZFKRBELIQ-UHFFFAOYSA-N
SMILESClC1=CC=C2OC(=O)NC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Formpowder
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only