1-Methylcyclopentanol, 98%
1-Methylcyclopentanol, 98%
1-Methylcyclopentanol, 98%
Thermo Scientific Chemicals

1-Methylcyclopentanol, 98%

CAS: 1462-03-9 | C6H12O | 100.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Catalog number B24744.06
also known as B24744-06
Price (EUR)
55,60
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
55,60
Each
Chemical Identifiers
CAS1462-03-9
IUPAC Name1-methylcyclopentan-1-ol
Molecular FormulaC6H12O
InChI KeyCAKWRXVKWGUISE-UHFFFAOYSA-N
SMILESCC1(O)CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormFused solid
Assay (GC)≥97.5%
Melting Point (clear melt)29.0-35.0?C
Water Content (Karl Fischer Titration)≤1%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only