Methyl 1,2,4-triazole-3-carboxylate, 98%
Methyl 1,2,4-triazole-3-carboxylate, 98%
Methyl 1,2,4-triazole-3-carboxylate, 98%
Thermo Scientific Chemicals

Methyl 1,2,4-triazole-3-carboxylate, 98%

CAS: 4928-88-5 | C4H5N3O2 | 127.103 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B24750.14
also known as B24750-14
Price (EUR)
133,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
133,00
Each
Chemical Identifiers
CAS4928-88-5
IUPAC Namemethyl 1H-1,2,4-triazole-5-carboxylate
Molecular FormulaC4H5N3O2
InChI KeyQMPFMODFBNEYJH-UHFFFAOYSA-N
SMILESCOC(=O)C1=NC=NN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder or flakes
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only