(2-Bromoethyl)triphenylphosphonium bromide, 98%
(2-Bromoethyl)triphenylphosphonium bromide, 98%
(2-Bromoethyl)triphenylphosphonium bromide, 98%
(2-Bromoethyl)triphenylphosphonium bromide, 98%
Thermo Scientific Chemicals

(2-Bromoethyl)triphenylphosphonium bromide, 98%

CAS: 7301-93-1 | C20H19Br2P | 450.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24833.14
also known as B24833-14
Price (EUR)
128,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
128,00
Each
Add to cart
Chemical Identifiers
CAS7301-93-1
IUPAC Name(2-bromoethyl)triphenylphosphanium bromide
Molecular FormulaC20H19Br2P
InChI KeyZHLVWYZWQPNQDQ-UHFFFAOYSA-M
SMILES[Br-].BrCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream
(2-Bromoethyl)triphenylphosphonium bromide is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(2-Bromoethyl)triphenylphosphonium bromide is used as pharmaceutical intermediates.

Notes
Hygroscopic. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
RUO – Research Use Only
Edward E. Schweizer, Robert D. Bach. Phosphonium Salts. II. 2-Bromophenetole and Triphenylphosphorus as Novel Phosphonioethylation Precursors. J. Org. Chem. 1964, 29, (7), 1746-1751.