1-Chloro-3,5-dimethoxybenzene, 98%
1-Chloro-3,5-dimethoxybenzene, 98%
1-Chloro-3,5-dimethoxybenzene, 98%
Thermo Scientific Chemicals

1-Chloro-3,5-dimethoxybenzene, 98%

CAS: 7051-16-3 | C8H9ClO2 | 172.608 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B24834.14
also known as B24834-14
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7051-16-3
IUPAC Name1-chloro-3,5-dimethoxybenzene
Molecular FormulaC8H9ClO2
InChI KeyWQHNWJBSROXROL-UHFFFAOYSA-N
SMILESCOC1=CC(OC)=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or fused solid or clear liquid as melt
Appearance (Color)White to yellow or pale cream to brown
Melting Point (clear melt)31.0-38.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only