Thermo Scientific Chemicals

Benzylurea, 98%, Thermo Scientific Chemicals

Catalog number: B24945.14
25 g, Each
Thermo Scientific Chemicals

Benzylurea, 98%, Thermo Scientific Chemicals

Catalog number: B24945.14
25 g, Each
Quantity
Catalog number: B24945.14
also known as B24945-14
Price (USD)
Price: 37.50
Online price: 31.65
Your price:
Quantity
-

Chemical Identifiers

CAS
538-32-9
IUPAC Name
benzylurea
Molecular Formula
C8H10N2O
InChI Key
RJNJWHFSKNJCTB-UHFFFAOYSA-N
SMILES
NC(=O)NCC1=CC=CC=C1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
147.0-153.0?C

Description

Benzylurea is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzylurea is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text