Thermo Scientific Chemicals

Methyl 2-hydroxy-3-nitrobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: B25029.06
5 g, Each
Thermo Scientific Chemicals

Methyl 2-hydroxy-3-nitrobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: B25029.06
5 g, Each
Quantity
Catalog number: B25029.06
also known as B25029-06
Price (USD)
Price: 158.00
Online price: 135.65
Your price:
Quantity
-

Chemical Identifiers

CAS
22621-41-6
IUPAC Name
2-(methoxycarbonyl)-6-nitrobenzen-1-olate
Molecular Formula
C8H6NO5
InChI Key
NIBVYEHAFBEVFI-UHFFFAOYSA-M
SMILES
COC(=O)C1=CC=CC(=C1[O-])[N+]([O-])=O
Appearance (Color)
Pale yellow to yellow to orange to pale brown
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
126.0-133.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text