Thermo Scientific Chemicals

Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals

Catalog number: B25049.36
500 g, Each
Thermo Scientific Chemicals

Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals

Catalog number: B25049.36
500 g, Each
Quantity
Catalog number: B25049.36
also known as B25049-36
Price (USD)
Price: 65.30
Online price: 55.65
Your price:
Quantity
-

Chemical Identifiers

CAS
97-97-2
IUPAC Name
2-chloro-1,1-dimethoxyethane
Molecular Formula
C4H9ClO2
InChI Key
CRZJPEIBPQWDGJ-UHFFFAOYSA-N
SMILES
COC(CCl)OC
Form
Liquid
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Refractive Index
1.4130-1.4170 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text