Thermo Scientific Chemicals

3,5-Dimethoxybenzyl alcohol, 99%, Thermo Scientific Chemicals

5g, Each
Thermo Scientific Chemicals

3,5-Dimethoxybenzyl alcohol, 99%, Thermo Scientific Chemicals

5g, Each
Quantity
Catalog number: B25138.06
also known as B25138-06
Price (USD)
Price: 85.70
Your Price:
Quantity
-

Chemical Identifiers

CAS
705-76-0
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%
Melting Point (clear melt)
44.5-50.5?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only