Thermo Scientific Chemicals

Glyoxylic acid, 50% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: B25149.0I
5000 g, Each
Thermo Scientific Chemicals

Glyoxylic acid, 50% w/w aq. soln., Thermo Scientific Chemicals

Catalog number: B25149.0I
5000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B25149.0I
also known as B25149-0I
Price (USD)
Price: 437.00
Online price: 371.65
Your price:
Quantity
-

Chemical Identifiers

CAS
298-12-4
IUPAC Name
2-oxoacetic acid
Molecular Formula
C2H2O3
InChI Key
HHLFWLYXYJOTON-UHFFFAOYSA-N
SMILES
OC(=O)C=O
Appearance (Color)
Clear colorless to yellow
Assay (Aqueous acid-base Titration)
≥50.0%
Refractive Index
1.4125-1.4200 @ 20?C
Form
Liquid

Description

Glyoxylic acid is used in Hopkins Cole reaction, which is used in the detection of tryptophan in proteins. It reacts with phenol to get 4-hydroxymandelic acid, which on further reaction with ammonia gives hydroxyphenylglycine, as a precursor to the drug amoxicillin. It is also used as a starting material for the preparation of 4-hydroxyphenylacetic acid, which is used to get atenolol. It is involved in the production of agrochemicals, aromas, cosmetic ingredient and pharmaceutical intermediate. It is also used in water purification and in the preservation of food. Further, it is employed as precursor in the synthesis of iron chelates. In addition to this, it serves as an intermediate of varnish material and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glyoxylic acid is used in Hopkins Cole reaction, which is used in the detection of tryptophan in proteins. It reacts with phenol to get 4-hydroxymandelic acid, which on further reaction with ammonia gives hydroxyphenylglycine, as a precursor to the drug amoxicillin. It is also used as a starting material for the preparation of 4-hydroxyphenylacetic acid, which is used to get atenolol. It is involved in the production of agrochemicals, aromas, cosmetic ingredient and pharmaceutical intermediate. It is also used in water purification and in the preservation of food. Further, it is employed as precursor in the synthesis of iron chelates. In addition to this, it serves as an intermediate of varnish material and dyes.

Solubility
Miscible with ethanol. Slightly miscible with ether and benzene. Immiscible with esters.

Notes
Incompatible with metals, alkalies, strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text