5-Bromo-2-fluorobenzylamine hydrochloride, 97%
5-Bromo-2-fluorobenzylamine hydrochloride, 97%
5-Bromo-2-fluorobenzylamine hydrochloride, 97%
Thermo Scientific Chemicals

5-Bromo-2-fluorobenzylamine hydrochloride, 97%

CAS: 202865-69-8 | C7H8BrFN | 205.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25176.14
also known as B25176-14
Price (EUR)
138,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
138,00
Each
Chemical Identifiers
CAS202865-69-8
IUPAC Name(5-bromo-2-fluorophenyl)methanaminium
Molecular FormulaC7H8BrFN
InChI KeyPDKBJZGGXHTHNC-UHFFFAOYSA-O
SMILES[NH3+]CC1=CC(Br)=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0%
Melting Point (clear melt)245.0-254.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only