(R)-(-)-2-Amino-1-butanol, 98%
(R)-(-)-2-Amino-1-butanol, 98%
(R)-(-)-2-Amino-1-butanol, 98%
Thermo Scientific Chemicals

(R)-(-)-2-Amino-1-butanol, 98%

CAS: 5856-63-3 | C4H11NO | 89.14 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) B25212.14
または、製品番号B25212-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5856-63-3
IUPAC Name2-aminobutan-1-ol
Molecular FormulaC4H11NO
InChI KeyJCBPETKZIGVZRE-UHFFFAOYNA-N
SMILESCCC(N)CO
さらに表示
Appearance (Color)Clear colorless to pale yellow
FormLiquid or viscous liquid
Assay (GC)≥97.5%
Water Content (Karl Fischer Titration)≤1%
Optical Rotation-6.5° to -10.5° (neat)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only