Ethyl 2-bromovalerate, 97%
Ethyl 2-bromovalerate, 97%
Ethyl 2-bromovalerate, 97%
Thermo Scientific Chemicals

Ethyl 2-bromovalerate, 97%

CAS: 615-83-8 | C7H13BrO2 | 209.083 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B25530.14
also known as B25530-14
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS615-83-8
IUPAC Nameethyl 2-bromopentanoate
Molecular FormulaC7H13BrO2
InChI KeyORSIRXYHFPHWTN-UHFFFAOYNA-N
SMILESCCCC(Br)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4470-1.4520 @ 20?C
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only