1-(1-Cyclohexen-1-yl)pyrrolidine, 97%
1-(1-Cyclohexen-1-yl)pyrrolidine, 97%
1-(1-Cyclohexen-1-yl)pyrrolidine, 97%
Thermo Scientific Chemicals

1-(1-Cyclohexen-1-yl)pyrrolidine, 97%

CAS: 1125-99-1 | C10H17N | 151.253 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B25622.06
also known as B25622-06
Price (EUR)
40,90
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
40,90
Each
Chemical Identifiers
CAS1125-99-1
IUPAC Name1-(cyclohex-1-en-1-yl)pyrrolidine
Molecular FormulaC10H17N
InChI KeyKTZNVZJECQAMBV-UHFFFAOYSA-N
SMILESC1CCN(C1)C1=CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to yellow
Assay (GC)>96.0%
Refractive Index1.5190-1.5240 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only