6-Bromo-1,4-benzodioxane, 98%
6-Bromo-1,4-benzodioxane, 98%
6-Bromo-1,4-benzodioxane, 98%
Thermo Scientific Chemicals

6-Bromo-1,4-benzodioxane, 98%

CAS: 52287-51-1 | C8H7BrO2 | 215.046 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25625.06
also known as B25625-06
Price (USD)
72.65
Online Exclusive
80.90
Save 8.25 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
72.65
Online Exclusive
80.90
Save 8.25 (10%)
Each
Chemical Identifiers
CAS52287-51-1
IUPAC Name6-bromo-2,3-dihydro-1,4-benzodioxine
Molecular FormulaC8H7BrO2
InChI KeyLFCURAJBHDNUNG-UHFFFAOYSA-N
SMILESBrC1=CC=C2OCCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5870-1.5910 @ 20?C
Appearance (Color)Clear colorless to pale yellow
6-Bromo-1,4-benzodioxane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromo-1,4-benzodioxane is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Séverine Jeulin; Sébastien Duprat de Paule; Virginie Ratovelomanana-Vidal; Jean-Pierre Genêt; Nicolas Champion and Philippe Dellis.Chiral biphenyl diphosphines for asymmetric catalysis: Stereoelectronic design and industrial perspectives. PNAS. 2004, 101 (16), 5799-5804.
  2. Dr. Lei Liang; Wei Wang; Jun Wu; Fengrong Xu; Dr. Yan Niu; Dr. Bo Xu; Dr. Ping Xu. Rational Design, Green Synthesis, and Initial Evaluation of a Series of Full-Color Tunable Fluorescent Dyes Enabled by the Copper-Catalyzed N-Arylation of 6-Phenyl Pyridazinones and Their Application in Cell Imaging. Chemistry A European Journal. 2013, 19 (41), 13774-13782.
  3. Lithiation can be effected at the 3-position using LDA or LTMP at -78°C: J. Org. Chem., 64, 8004 (1999).